Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N4BP2L2 Antibody (OTI1C8), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP246078
Description
Ensure accurate, reproducible results in Western Blot, Immunohistochemistry
N4BP2L2 Monoclonal specifically detects N4BP2L2 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
N4BP2L2 | |
Monoclonal | |
1 mg/mL | |
Western Blot 1:200, Immunohistochemistry 1:150 | |
CG005FLJ41089, CG016, FLJ36195, NEDD4 binding protein 2-like 2, NEDD4-binding protein 2-like 2, PFAAP5FLJ43077, Phosphonoformate immuno-associated protein 5,92M18.3,92M18.3 (novel protein), protein from BCRA2 region | |
Mouse | |
67.3 kDa | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
OTI1C8 | |
Unconjugated | |
PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide | |
N4BP2L2 | |
Full length human recombinant protein of human N4BP2L2 (NP_055702) produced in E.coli. | |
Immunogen affinity purified | |
RUO | |
10443 | |
Store at -20C. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction