Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N4BP2L2 Monoclonal Antibody (OTI1C8)
GREENER_CHOICE

N4BP2L2 Monoclonal Antibody (OTI1C8)

Supplier:  Thermo Scientific MA526655

Catalog No. PIMA526655


Only null left
Add to Cart

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

N4BP2L2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q92802
N4BP2L2
Full length human recombinant protein of N4BP2L2 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG2a
Immunohistochemistry (Paraffin), Western Blot
OTI1C8
Unconjugated
N4BP2L2
2700092H06Rik; 92M18.3; CG005; CG016; N4BP2L2; NEDD4 binding protein 2 like 2; NEDD4 binding protein 2-like 2; NEDD4-binding protein 2-like 2; PFAAP5; phosphonoformate immuno-associated protein 5; Phosphonoformate immuno-associated protein 5 homolog; protein from BRCA2 region; zag1
Mouse
Affinity Chromatography
RUO
10443
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.