Learn More
N4BP2L2 Mouse anti-Human, Clone: OTI5A3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF804200

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
N4BP2L2 (NEDD4-binding protein 2-like 2), also known as PFAAP5 (phosphonoformate immuno-associated protein 5), is a 583 amino acid nuclear protein that potentially is involved in transcriptional regulation. N4BP2L2 is phosphorylated on Ser 199 in response to DNA damage, probably by ATM or ATR. Primarily expressed in bone marrow, N4BP2L2 is dramatically down-regulated after exposure to arsenic compounds, an event which precedes neutropenia. PFAAP5 interacts with both Gfi-1 and Neutrophil Elastase, two proteins that are implicated in neutropenia disorders. Defects in the gene encoding Neutrophil Elastase, ELA2, are the cause of cyclic haematopoiesis, which, with decreased numbers of circulating neutrophils, leads to an increased risk for opportunistic infection.Specifications
N4BP2L2 | |
Monoclonal | |
Unconjugated | |
N4BP2L2 | |
92M18.3, CG005, CG016, PFAAP5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10443 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI5A3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q92802 | |
N4BP2L2 | |
Full length human recombit protein of human N4BP2L2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.