Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ N4BP3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556606 Supplier Invitrogen™ Supplier No. PA556606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: HLNHLGGSLD RASQGPKEAG PPAVLSCLPE PPPPYEFSCS SAEEMGAVLP ETCEELKRGL GDEDGSNPFT QVLEERQRLW LAELKRLYVE RLHE Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 83%.

N4BP3, also named as KIAA0341, a Fezzin family member, during nerve cell development. It is crucial for axonal and dendritic branching in developing neurons. It binds the protein NEDD4 and interacts with 14-3-3 proteins. N4BP3 is present in early neurites, growth cones, axons and dendrites of developing primary hippocampal neurons.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N4BP3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene N4BP3
Gene Accession No. O15049
Gene Alias C330016O10Rik; EMBL:ABA06436.1}; KIAA0341; LZTS4; N4bp3; N4BP3 {ECO:0000250; N4bp3 {ECO:0000312; Nedd4 binding protein 3; nedd4 binding protein 3 {ECO:0000312; NEDD4-binding protein 3; RGD:1308356}; RGD1308356; UniProtKB:O15049}
Gene Symbols N4BP3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human N4BP3. Recombinant protein control fragment (Product #RP-104173).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23138
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.