Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ N4BP3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA556607
Description
Immunogen sequence: FSPEELRAAL AGSRGSRQPD GLLRKGLGQR EFLSYLHLPK KDSKSTKNTK RAPRNEPADY ATLYYREHSR AGDFSKTSLP ERGRFD Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 88%.
N4BP3, also named as KIAA0341, a Fezzin family member, during nerve cell development. It is crucial for axonal and dendritic branching in developing neurons. It binds the protein NEDD4 and interacts with 14-3-3 proteins. N4BP3 is present in early neurites, growth cones, axons and dendrites of developing primary hippocampal neurons.
Specifications
| N4BP3 | |
| Polyclonal | |
| Unconjugated | |
| N4BP3 | |
| C330016O10Rik; EMBL:ABA06436.1}; KIAA0341; LZTS4; N4bp3; N4BP3 {ECO:0000250; N4bp3 {ECO:0000312; Nedd4 binding protein 3; nedd4 binding protein 3 {ECO:0000312; NEDD4-binding protein 3; RGD:1308356}; RGD1308356; UniProtKB:O15049} | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23138 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.8 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O15049 | |
| N4BP3 | |
| Recombinant protein corresponding to Human N4BP3. Recombinant protein control fragment (Product #RP-95516). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction