Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N6-methyladenosine (m6A) Antibody (17-3-4-1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP250525R
Description
N6-methyladenosine (m6A) Monoclonal specifically detects N6-methyladenosine (m6A) in Human, Mouse, Yeast samples. It is validated for Western Blot.Specifications
N6-methyladenosine (m6A) | |
Monoclonal | |
DyLight 550 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Hapten N6-methyladenosine-5'-mono-phosphate conjugated to BSA of all N6-methyladenosine | |
0.1 mL | |
Primary | |
Human, Mouse, Yeast | |
IgG1 λ |
Western Blot | |
17-3-4-1 | |
Western Blot | |
Mouse | |
Protein A purified | |
Epigenetics, metabolism | |
This antibody recognizes N6-methyladenosine in both modified RNA AND DNA. | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction