Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
N6-Methyladenosine (m6A) Monoclonal Antibody (17-3-4-1), Active Motif™

Mouse Monoclonal Antibody
$156.65 - $620.58
Specifications
Antigen | N6-Methyladenosine (m6A) |
---|---|
Clone | 17-3-4-1 |
Concentration | 1 mg/mL |
Applications | Dot Blot, Immunoprecipitation |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3674
![]() ![]() |
Active Motif
61756 |
10 μg |
Each for $156.65
|
|
|||||
50-199-3673
![]() ![]() |
Active Motif
61755 |
100 μg |
Each for $620.58
|
|
|||||
Description
N6-methyladenosine (m6A), or methylation of the N6 position of adenosine is a posttranscriptional modification of RNA. Due to a lack of analytical methods, N6-methyladenosine is poorly understood, but new evidence suggests that it is a very common base modification and important physiological regulator. N6-methyladenosine is markedly increased throughout brain development, and is enriched near stop codons, microRNA-binding sites and UTRs, which indicates a fundamental role in the regulation of gene expression. N6-methyladenosine is also highly conserved between human and mouse.Specifications
N6-Methyladenosine (m6A) | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 | |
IgG1 κ | |
Protein A | |
Antibody |
17-3-4-1 | |
Dot Blot, Immunoprecipitation | |
Unconjugated | |
Mouse | |
Human, Yeast | |
This antibody was raised against 6-Methyladenosine conjugated to BSA. | |
Primary | |
-20°C, Avoid Freeze/Thaw Cycles |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title