Learn More
N6-Methyladenosine (m6A) Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | N6-Methyladenosine (m6A) |
| Applications | Dot Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Host Species | Rabbit |
| Immunogen | This antibody was raised against 6-Methyladenosine conjugated to kLH. |
| Purification Method | Protein A |
| Quantity | 10 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.