Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N6-Methyladenosine Rabbit anti-Human, Unconjugated, Clone: RM362, Abnova™

Catalog No. 89339212 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-212 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-212 Supplier Abnova Corporation Supplier No. MAB21995
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody.

N6-Methyladenosine Monoclonal antibody specifically detects N6-Methyladenosine in Human samples. It is validated for ELISA, Dot-Pep, RIP-A.

Specifications

Antigen N6-Methyladenosine
Applications ELISA, Dot Blot, RIP Assay
Classification Monoclonal
Clone RM362
Conjugate Unconjugated
Host Species Rabbit
Immunogen BSA-conjugated N6-Methyladenosine
Quantity 50 μg
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.