Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ N6AMT1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566242
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566242 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566242 Supplier Invitrogen™ Supplier No. PA566242
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GREVMDRFFPL VPDLLSPRGL FYLVTIKENN PEEILKIMKT KGLQGTTALS RQAGQETLSV LKFT Highest antigen sequence identity to the following orthologs - mouse 85%, rat 88%.

Heterodimeric methyltransferase that catalyzes N5-methylation of ETF1 on 'Gln-185', using S-adenosyl L-methionine as methyl donor. ETF1 needs to be complexed to ERF3 in its GTP-bound form to be efficiently methylated. May play a role in the modulation of arsenic-induced toxicity. May be involved in the conversion of monomethylarsonous acid (3+) into the less toxic dimethylarsonic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N6AMT1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene N6AMT1
Gene Accession No. Q9Y5N5
Gene Alias 5830445C04Rik; C21orf127; HemK methyltransferase family member 2; HEMK2; m.HsaHemK2P; Methylarsonite methyltransferase N6AMT1; Methyltransferase N6AMT1; methyltransferase N6AMT1; hemK methyltransferase family member 2; M-N6DNA MTase A; MTQ2; N(6)-adenine-specific DNA methyltransferase 1; N(6)-adenine-specific DNA methyltransferase 1-like; N-6 adenine-specific DNA methyltransferase 1; N-6 adenine-specific DNA methyltransferase 1 (putative); N6AMT; N6amt1; N6-DNA methyltransferase A; N6-DNA-methyltransferase; PRED28; Protein N(5)-glutamine methyltransferase; RGD1311843
Gene Symbols N6AMT1
Host Species Rabbit
Immunogen Recombinant Human N6AMT1. Recombinant protein control fragment (Product #RP-106922).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29104
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.