Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ N6AMT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA560017
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA560017 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA560017 Supplier Invitrogen™ Supplier No. PA560017
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GEGGRIACVS APSVYQKLRE LCRENFSIYI FEYDKRFAMY GEEFIFYDYN NPLDLPERIA AHSFDIVIAD PPY Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 85%.

Methylation of DNA at cytosine residues plays an important role in regulation of gene expression, genomic imprinting and is essential for mammalian development. Hypermethylation of CpG islands in tumor suppressor genes or hypomethylation of bulk genomic DNA may be linked with development of cancer. To date, 3 families of mammalian DNA methyltransferase genes have been identified which include Dnmt1, Dnmt2 and Dnmt3. Dnmt1 is constitutively expressed in proliferating cells and inactivation of this gene causes global demethylation of genomic DNA and embryonic lethality. Dnmt2 is expressed at low levels in adult tissues and its inactivation does not affect DNA methylation or maintenance of methylation. Dnmt2 contains all the sequence motifs diagnostic of DNA (cytosine-5)-methyltransferases but appears to lack the large N-terminal regulatory domain common to other eukaryotic methyltransferases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen N6AMT2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Eef1akmt1
Gene Accession No. Q8WVE0
Gene Alias 2510005D08Rik; AW045965; Ayu21-96; DMNT2; DNMT2; EEF1A lysine methyltransferase 1; Eef1akmt1; ESP13; eukaryotic translation elongation factor 1 alpha lysine methyltransferase 1; Gt(Ayu21)96Imeg; Gt(pU21)96Imeg; GtAyu21-96; M.HsaIIP; N(6)-adenine-specific DNA methyltransferase 2; N-6 adenine-specific DNA methyltransferase 2 (putative); N6AMT2; Protein-lysine N-methyltransferase N6AMT2; PuMet; RGD1306762; RNMT1; RP11-406H21.1
Gene Symbols Eef1akmt1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human N6AMT2. Recombinant protein control fragment (Product #RP-97984).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 221143
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.