Learn More
3M™ N95 Particulate Respirator with Nuisance Level Organic Vapor Relief†
Catalog No. p-7237793 Shop All 3M Products
Change view
Click to view available options
With Faceseal:
No
Yes
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. With Faceseal
19-034-803 Yes
19-037059 No
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 19-034-803 Supplier 3M™ Supplier No. 7000002083
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
NIOSH approved respirator used for nuisance level organic vapors such as solvents, degreasers, and/or resins.
  • NIOSH approved N95 rating
  • Cake resistant filter media
  • Foam faceseal
  • Adjustable buckle straps
  • Flame resistant per modified ASTM D2859
  • Cool Flow™ Exhalation Valve
  • Carbon layer for ozone/nuisance level organic vapors†
  • † Nuisance level refers to concentrations not exceeding OSHA PEL or applicable occupational exposure limit whichever is lower
TRUSTED_SUSTAINABILITY
Specifications
Aerosol Type N - Not Resistant to Oil
Assigned Protection Factor (APF) 10
Certifications/Compliance ASTM D2859
Certifications (NIOSH) NIOSH 42 CFR 84, N95, approval number: TC-84A-0454
Certifications (OSHA) OSHA 1910.134
Color Code White
Design Type Cup/Molded
Disposable Single-use
Filter Type N95
Material Polypropylene, Polyester
Product Type Particulate Respirator with Nuisance Organic Vapor Relief
Size Standard
With Exhalation Valve Yes
Filter Efficiency 95%
For Use With (Application) Assembly and mechanical, cleaning, composite finishing, foundry operations, grinding, paint preparation, petrochemical, processing of minerals, welding
Latex Free Yes
Material Headstrap Braided Polyisoprene, Cotton/Polyester
Model 8214
Nose Piece Material Aluminum
Nosepiece Type Standard
Nuisance Odor Relief Organic Vapors
Quantity 80/Cs.
Sterility Non-sterile
With Faceseal Yes
Strap Attachment Type Adjustable Buckle Straps
FDA Cleared Surgical Mask No