Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NA-XTD™ Influenza Neuraminidase Assay Kit
GREENER_CHOICE

Catalog No. 4457535
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 x 96 well plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-575-35 10 x 96 well plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 44-575-35 Supplier Invitrogen™ Supplier No. 4457535
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Includes

Influenza Neuraminidase Assay Kit includes 1000X NA-XTD Substrate, NA Sample Prep Buffer, NA-XTD Assay Buffer, NA-XTD Accelerator and 10 detection microplates Influenza Neuraminidase Assay Reagent Set includes 1000X NA-XTD Substrate, NA Sample Prep Buffer

Next-generation chemiluminescence-based assay for long-lived emission kinetics for quantitation of influenza neuraminidase inhibitor resistance. Thermo Scientific NA-XTD Influenza Neuraminidase Assay Kit can be used for virus quantitation in media samples

The NA-XTD™ Influenza Neuraminidase Assay Kit is a next-generation chemiluminescence-based assay that provides a longer signal read-out compared to the first-generation NA-Star® Influenza Neuraminidase Inhibitor Resistance Detection Kit. The NA-XTD™ kit includes detection reagents and microplates, eliminating the need for luminometers equipped with a reagent injector, and improving ease-of-use. This kit also includes complete assay protocols for quantitating sensitivity to neuraminidase inhibitors in various influenza virus isolates including: human influenza types A and B, A/H1N1 pandemic, avian, equine and porcine viruses.

Key product features:

  • Superior performance—improved, long-lived light emission kinetics and read-time flexibility
  • High sensitivity—high assay signal-to-noise, detection of low-virus concentrations, and wide assay dynamic range
  • Simple instrumentation requirement—no reagent injectors or luminometer needed
  • Multiple applications—neuraminidase inhibitor IC50 assays and cell-based virus quantitation

Long-Lived Light Emission Kinetics & Read-Time Flexibility

The NA-XTD™ assay yields much longer-lived light emission kinetics than does the NA-Star® assay, eliminating the need for luminometer instrumentation with reagent injectors and enabling read-time flexibility and batch-mode processing of assay plates. NA-XTD™ assay signal half-life is approximately 2 hours, longer than the 5-10 minute half-life of the NA-Star® assay signal. IC-50 values determined from data collected immediately or up to 3 hours following addition of NA-XTD™ Accelerator solution are identical (Figure 1).

High Sensitivity

The NA-XTD™ chemiluminescent assay provides higher detection sensitivity (better low-end detection limit), higher assay signal-to-noise ratio, and wider assay dynamic range than fluorescent assays with the MUNANA substrate. The NA-XTD™ assay also typically demonstrates slightly higher signal-to-noise than the NA-Star® assay. The NA-XTD™ assay provides 2–50-fold higher sensitivity by signal-to-noise ratio than MUNANA-based fluorescence assays, depending on the virus isolate (Figure 2). The NA-XTD™ assay provides a dynamic range of detection of 3–4 orders of magnitude of neuraminidase enzyme concentration, compared to 2–3 orders of magnitude range with fluorescent MUNANA assays. The wide chemiluminescent neuraminidase assay range enables determination of IC-50 values over a wide range of virus concentrations, eliminating the need to titer virus prior to performing IC50 determination assays (Figure 3).

Simple Instrumentation Requirement

The NA-XTD ™ assay can be used for virus quantitation in media samples from 96-well microplates or other virus cultures for monitoring viral growth or infection, or for performing viral inhibition assays in a cell-based system. Optimally, a small sample of culture media is removed and assayed directly with NA-XTD™ reagents, permitting multiple samples to be assayed over time (Figure 4).

Multiple Applications in One Complete Kit

The NA-XTD™ Assay Buffer is used as diluent for virus samples, neuraminidase inhibitors and NA-XTD™ substrate. An optional NA Sample Prep Buffer is also included for Triton® X-100 detergent addition to virus preparations, which increases NA activity in some virus preparations. The next-generation NA-XTD™ Accelerator solution triggers high intensity light emission from the NA-XTD™ reaction product. For added convenience and assay performance, the kit includes NA-Star™ Detection Microplates. These 96-well solid white assay microplates were selected for optimum assay performance, including high signal intensity, low background, and minimum well-to-well cross-talk. The kit includes a comprehensive assay protocol that provides virus and neuraminidase inhibitor (NI) dilution recommendations, a recommended plate layout for NI sensitivity assays, a protocol for virus quantitation, and a literature reference list. The NA-XTD™ Influenza Neuraminidase Assay Kit is compatible with a wide range of luminometers, including single-mode and multi-mode instruments, with no need for injectors.

For Research Use Only. Not for use in diagnostics procedures.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage This kit contains 1000X NA-XTD™ Substrate, NA Sample Prep Buffer, NA-XTD™ Assay Buffer, NA-XTD™ Accelerator, and 10 detection microplates. Store all reagents at 2°C to 8°C.
Detection Method Luminescence
For Use With (Application) Neuraminidase (Sialidase) Assay
For Use With (Equipment) Luminometer
Product Type Neuraminidase Assay
Form Liquid
Format 96-well plate
Product Line NA-XTD, NovaBright
Quantity 10 x 96 well plates
Substrate NA-XTD™
Substrate Properties Chemical Substrate
Substrate Type Neuraminidase Substrate
Target Enzyme Sialidase, Neuraminidase
Throughput High-throughput Compatible
Validated Application Enzyme Assay
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.