Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAA10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
Antigen | NAA10 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Description
NAA10 Polyclonal specifically detects NAA10 in Human samples. It is validated for Western Blot.Specifications
NAA10 | |
Polyclonal | |
Rabbit | |
P41227 | |
8260 | |
Synthetic peptides corresponding to NAA10 N-acetyltransferase (S. cerevisiae)) The peptide sequence was selected from the middle region of ARD1A. Peptide sequence VESKGNSPPSSGEACREEKGLAAEDSGGDSKDLSEVSETTESTDVKDSSE. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
ARD1, ARD1 homolog A, N-acetyltransferase (S. cerevisiae), ARD1 homolog, N-acetyltransferase (S. cerevisiae), ARD1AARD1 homolog, N-acetyltransferase, EC 2.3.1.-, EC 2.3.1.88, MGC71248, N(alpha)-acetyltransferase 10, NatA catalytic subunit, N-acetyltransferase, N-acetyltransferase ARD1, human homolog of, N-alpha-acetyltransferase 10, NatA catalytic subunit, N-terminal acetyltransferase complex ARD1 subunit homolog A | |
NAA10 | |
IgG | |
26 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title