Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAA15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA575957
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA575957 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA575957 Supplier Invitrogen™ Supplier No. PA575957
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

This gene encodes a protein of unknown function. However, similarity to proteins in yeast and other species suggests that this protein may be an N-acetyltransferase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAA15
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NAA15
Gene Accession No. Q80UM3, Q9BXJ9
Gene Alias 5730450D16Rik; 6330400I15; ASTBDN; GA19; gastric cancer antigen Ga19; mNAT1; N(alpha)-acetyltransferase 15, NatA auxiliary subunit; Naa15; N-alpha-acetyltransferase 15, NatA auxiliary subunit; NARG1; NAT1P; NATH; NMDA receptor regulated 1; NMDA receptor-regulated gene 1; NMDA receptor-regulated protein 1; N-terminal acetyltransferase; N-terminal acetyltransferase 1; N-terminal aceyltransferase 1; Protein tubedown-1; TBDN; Tbdn-1; TBDN100; transcriptional coactivator tubedown-100; tubedown; tubedown-1
Gene Symbols NAA15
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 225-269 of Human NAA15.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310399, 74838, 80155
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.