Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAA16 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA558917
Description
Immunogen sequence: FAINSNNPWL HECLIRFSKS VSNHSNLPDI VSKVLSQEMQ KIFVKKDLES FNEDFLKRNA TSLQHLLSGA KMMYFLDKSR QEKA Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 88%.
NARG1L may belong to a complex displaying N-terminal acetyltransferase activity.
Specifications
NAA16 | |
Polyclonal | |
Unconjugated | |
NAA16 | |
1300019C06Rik; N(alpha)-acetyltransferase 16, NatA auxiliary subunit; NAA16; Nalpha acetyltransferase 16; N-alpha-acetyltransferase 16, NatA auxiliary subunit; Narg1l; NARG1-like protein; NAT2; NMDA receptor regulated 1-like protein; NMDA receptor-regulated 1-like protein; NMDA receptor-regulated 1-like protein-like protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
79612 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q6N069 | |
NAA16 | |
Recombinant protein corresponding to Human NAA16. Recombinant protein control fragment (Product #RP-97991). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction