Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAA20 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596579
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596579 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596579 Supplier Invitrogen™ Supplier No. PA596579
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PGGELMGYIM GKAEGSVARE EWHGHVTALS VAPEFRRLGL AAKLMELLEE ISERKGGFFV DLFVRVSNQV AVNMYKQLGY SVYRTVIEYY SASNGEPDED AYDMRKALSR DTEKKSIIPL PHPVRPEDIE; Positive Samples: HeLa, Mouse brain, Mouse kidney, Rat brain; Cellular Location: Cytoplasm, Nucleus.

Catalytic subunit of the NatB complex which catalyzes acetylation of the N-terminal methionine residues of peptides beginning with Met-Asp, Met-Glu, Met-Asn and Met-Gln. Proteins with cell cycle functions are overrepresented in the pool of NatB substrates. Required for maintaining the structure and function of actomyosin fibers and for proper cellular migration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAA20
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.31 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene NAA20
Gene Accession No. P61599, P61600
Gene Alias 1500004D14Rik; 2900026I01Rik; AU041458; D2Ertd186e; dJ1002M8.1; Methionine N-acetyltransferase; N(alpha)-acetyltransferase 20, NatB catalytic subunit; NAA20; N-acetyltransferase 3 homolog; N-acetyltransferase 5; N-acetyltransferase 5 (ARD1 homolog, S. cerevisiae); N-acetyltransferase 5 (GCN5-related, putative); N-acetyltransferase 5, ARD1 subunit (arrest-defective 1, S. cerevisiae, homolog); N-alpha-acetyltransferase 20; N-alpha-acetyltransferase 20, NatB catalytic subunit; NAT3; NAT3P; NAT5; NAT5P; NatB catalytic subunit; natB complex subunit NAT5; N-terminal acetyltransferase B complex catalytic subunit NAA20; N-terminal acetyltransferase B complex catalytic subunit NAT5; N-terminal acetyltransferase complex ARD1 subunit
Gene Symbols NAA20
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 49-178 of human NAA20 (NP_0571841).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 362228, 51126, 67877
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.