Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAA30 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5107003
Description
Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 91%; Pig: 100%; Rabbit: 93%; Rat: 91%.
Catalytic subunit of the N-terminal acetyltransferase C (NatC) complex. Catalyzes acetylation of the N-terminal methionine residues of peptides beginning with Met-Leu-Ala and Met-Leu-Gly. Necessary for the lysosomal localization and function of ARL8B sugeesting that ARL8B is a NatC substrate.
Specifications
| NAA30 | |
| Polyclonal | |
| Unconjugated | |
| NAA30 | |
| 4930487N19Rik; 5730533P17Rik; AI447922; AW322455; C14orf35; MAK3; Mak3p; N(alpha)-acetyltransferase 30, NatC catalytic subunit; NAA30; N-acetyltransferase 12; N-acetyltransferase 12 (GCN5-related, putative); N-acetyltransferase MAK3 homolog; Nalpha acetyltransferase 30; N-alpha-acetyltransferase 30; N-alpha-acetyltransferase 30, NatC catalytic subunit; NAT12; NAT12P; NatC catalytic subunit; putative N-acetyltransferase | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 122830 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide; pH 7.4 | |
| Q147X3 | |
| NAA30 | |
| Synthetic peptide directed towards the middle region of human NAT12 (aa 182-232). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction