Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAA38 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA570485
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
NAA38 is a protein coding gene.
Specifications
NAA38 | |
Polyclonal | |
Unconjugated | |
NAA38 | |
1500034E06Rik; fj33f12; LSM domain containing 1; LSM domain-containing protein 1; LSMD1; MAK31; MAK31-like protein; N(alpha)-acetyltransferase 38, NatC auxiliary subunit; naa38; N-alpha-acetyltransferase 38, NatC auxiliary subunit; PFAAP2; Phosphonoformate immuno-associated protein 2; RGD1559617; si:ch211-30g3.3; wu:fj33f12 | |
Rabbit | |
Affinity chromatography | |
RUO | |
84316 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q9BRA0 | |
NAA38 | |
synthetic peptide directed towards the C-terminal region of human NAA38. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction