Learn More
Invitrogen™ NAA38 Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 80%; Dog: 86%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Mouse: 79%; Rabbit: 86%; Rat: 79%.
Specifications
Specifications
| Antigen | NAA38 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | NAA38 |
| Gene Accession No. | Q9BRA0 |
| Gene Alias | 1500034E06Rik; fj33f12; LSM domain containing 1; LSM domain-containing protein 1; LSMD1; MAK31; MAK31-like protein; N(alpha)-acetyltransferase 38, NatC auxiliary subunit; naa38; N-alpha-acetyltransferase 38, NatC auxiliary subunit; PFAAP2; Phosphonoformate immuno-associated protein 2; RGD1559617; si:ch211-30g3.3; wu:fj33f12 |
| Gene Symbols | NAA38 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.