Learn More
NAA50 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305594AT

Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine and Orangutan
NAT13 is a probable catalytic component of the ARD1A-NARG1 complex which displays alpha (N-terminal) acetyltransferase activity.Specifications
NAA50 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9GZZ1 | |
NAA50 | |
Between 119 and 169 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Immunoprecipitation | |
Unconjugated | |
NAA50 | |
MAK3, NAT13, NAT5, SAN, hNAT5, hSAN, Mak3 homolog, N-acetyltransferase 13 (GCN5-related), N-acetyltransferase 5, N-acetyltransferase san homolog, N-alpha-acetyltransferase 50, N-alpha-acetyltransferase 50, NatE catalytic subunit, natE catalytic subunit, separation anxiety | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80218 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.