Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ NAADP tetrasodium salt
GREENER_CHOICESDP

Catalog No. 390510 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
39-051-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 39-051-0 Supplier R&D Systems™ Supplier No. 3905/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Ca2+ mobilizing agent

Ca2+ mobilizing agent. Initiates Ca2+ release via type 1 ryanodine receptor (RyR1) activation. Also regulates Ca2+ release from intracellular stores distinct from the endoplasmic reticulum (ER); displays affinity for two-pore channels (TPCs) which release Ca2+ from acidic organelles.

Chemical Identifiers

CAS 5502-96-5
Molecular Formula C21H23N6Na4O18P3

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 832.32
Purity >98%
Molecular Weight (g/mol) 832.319
Chemical Name or Material NAADP tetrasodium salt
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.