Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAALAD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA566773
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA566773 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA566773 Supplier Invitrogen™ Supplier No. PA566773
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NDAEILLRYLG GIAPPDKSWK GALNVSYSIG PGFTGSDSFR KVRMHVYNIN Highest antigen sequence identity to the following orthologs - mouse 78%, rat 78%.

Has N-acetylated-alpha-linked-acidic dipeptidase (NAALADase) activity. Also exhibits a dipeptidyl-peptidase IV type activity. Inactivate the peptide neurotransmitter N- acetylaspartylglutamate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAALAD2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NAALAD2
Gene Accession No. Q9Y3Q0
Gene Alias GCPIII; Glutamate carboxypeptidase III; GPCIII; NAADALASE2; NAALAD2; NAALADase II; NAALADASE2; N-acetylated alpha-linked acidic dipeptidase 2; N-acetylated alpha-linked acidic dipeptidase II; N-acetylated-alpha-linked acidic dipeptidase 2; N-acetylated-alpha-linked acidic dipeptidase II
Gene Symbols NAALAD2
Host Species Rabbit
Immunogen Recombinant Human NAALAD2. Recombinant protein control fragment (Product #RP-107430).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10003
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.