Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAALADase-like 1/NAALADL1 Antibody (026), Alexa Fluor™ 750, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP290749AF750
Description
NAALADase-like 1/NAALADL1 Monoclonal specifically detects NAALADase-like 1/NAALADL1 in Mouse samples. It is validated for ELISA.Specifications
NAALADase-like 1/NAALADL1 | |
Monoclonal | |
Alexa Fluor 750 | |
I100, NAALADASEL, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1 | |
This antibody was obtained from a rabbit immunized with purified, recombinant Mouse NAALADase-like 1/NAALADL1 (Q7M758; Pro29-Leu745). | |
0.1 mL | |
Primary | |
No cross-reactivity in ELISA with: Human NAALADL1 | |
Store at 4°C in the dark. | |
IgG |
ELISA | |
26 | |
50 mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
10004 | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction