Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAALADase-like 1/NAALADL1 Antibody, mFluor Violet 610 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299724MFV610
Description
NAALADase-like 1/NAALADL1 Polyclonal antibody specifically detects NAALADase-like 1/NAALADL1 in Human samples. It is validated for ELISASpecifications
NAALADase-like 1/NAALADL1 | |
Polyclonal | |
ELISA | |
I100, NAALADASEL, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1 | |
Produced in rabbits immunized with purified, recombinant Human NAALADase-like 1/NAALADL1 (Accession#: NP_005459.2; Pro 29-Leu 740) | |
0.1 mL | |
Primary | |
Human | |
Purified |
ELISA | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
10004 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction