Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAALADase-like 1/NAALADL1 Rabbit anti-Human, Clone: 29, Novus Biologicals™

Rabbit Monoclonal Antibody
$353.50 - $560.50
Specifications
Antigen | NAALADase-like 1/NAALADL1 |
---|---|
Clone | 29 |
Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 |
Applications | Western Blot, ELISA |
Classification | Monoclonal |
Description
NAALADase-like 1/NAALADL1 Monoclonal specifically detects NAALADase-like 1/NAALADL1 in Human samples. It is validated for Western Blot, ELISA.Specifications
NAALADase-like 1/NAALADL1 | |
Western Blot 1:500-1:1000, ELISA 1:25000-1:50000 | |
Monoclonal | |
Purified | |
RUO | |
0.2 um filtered solution in PBS | |
10004 | |
IgG | |
Protein A purified |
29 | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
Human | |
I100, NAALADASEL, NAALADL1 N-acetylated alpha-linked acidic dipeptidase-like 1 | |
Recombinant Human NAALADase-like 1/NAALADL1 protein | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title