missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NAALADase-like 2/NAALADL2 Antibody (817225), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4665Z
Description
NAALADase-like 2/NAALADL2 Monoclonal antibody specifically detects NAALADase-like 2/NAALADL2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
NAALADase-like 2/NAALADL2 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
254827 | |
Store at 4°C in the dark. | |
IgG1 |
Flow Cytometry, CyTOF | |
817225 | |
Flow Cytometry, CyTOF-ready | |
inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2, NAALADase L2, N-acetylated alpha-linked acidic dipeptidase 2, N-acetylated alpha-linked acidic dipeptidase-like 2 | |
Mouse myeloma cell line NS0-derived recombinant human NAALADase-like 2/NAALADL2, Ser152-Asn795, Accession # Q58DX5 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction