Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAALADase-like 2/NAALADL2 Antibody (817225), mFluor Violet 610 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4665MFV610
Description
NAALADase-like 2/NAALADL2 Monoclonal antibody specifically detects NAALADase-like 2/NAALADL2 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
| NAALADase-like 2/NAALADL2 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 254827 | |
| Store at 4C in the dark. | |
| IgG1 |
| Flow Cytometry, CyTOF | |
| 817225 | |
| Flow Cytometry, CyTOF-ready | |
| inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2, NAALADase L2, N-acetylated alpha-linked acidic dipeptidase 2, N-acetylated alpha-linked acidic dipeptidase-like 2 | |
| Mouse myeloma cell line NS0-derived recombinant human NAALADase-like 2/NAALADL2, Ser152-Asn795, Accession # Q58DX5 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction