Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ NAALADL1 Monoclonal Antibody (3C5G10E8)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA530690
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human NAALADL1/NAALADase L.
NAALADL1 has NAALADase-like activity which is currenty unknown in function. NAALADL1 has no NAAG hydrolyzing activity, exhibits a dipeptidyl-peptidase IV type activity. In vitro, cleaves Gly-Pro-AMC.
Specifications
NAALADL1 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative | |
Q9UQQ1 | |
Naaladl1 | |
Recombinant Human NAALADL1 protein (Pro29-Leu740). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
ELISA | |
3C5G10E8 | |
Unconjugated | |
Naaladl1 | |
100 kDa ileum brush border membrane protein; Aminopeptidase NAALADL1; Gm964; I100; ileal dipeptidylpeptidase; ileal peptidase I100; NAALADase L; NAALADASEL; NAALADase-like 1; NAALADL; Naaladl1; N-acetylated alpha-linked acidic dipeptidase-like 1; N-acetylated-alpha-linked acidic dipeptidase like protein; N-acetylated-alpha-linked acidic dipeptidase-like protein | |
Mouse | |
Protein A | |
RUO | |
10004 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction