Learn More
Invitrogen™ NAALADL1 Polyclonal Antibody

Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Mouse NAALADL1.
Specifications
Specifications
| Antigen | NAALADL1 |
| Applications | ELISA, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene | Naaladl1 |
| Gene Accession No. | Q7M758 |
| Gene Alias | 100 kDa ileum brush border membrane protein; Aminopeptidase NAALADL1; Gm964; I100; ileal dipeptidylpeptidase; ileal peptidase I100; NAALADase L; NAALADASEL; NAALADase-like 1; NAALADL; Naaladl1; N-acetylated alpha-linked acidic dipeptidase-like 1; N-acetylated-alpha-linked acidic dipeptidase like protein; N-acetylated-alpha-linked acidic dipeptidase-like protein |
| Gene Symbols | Naaladl1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.