Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAALADL2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547647
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547647 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547647 Supplier Invitrogen™ Supplier No. PA547647
Add to Cart
Add to Cart

Goat Polyclonal Antibody

In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse NAALADase 2 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

NAALADL2 (N-Acetylated Alpha-Linked Acidic Dipeptidase Like 2) is a Protein Coding gene and may be catalytically inactive. Diseases associated with NAALADL2 include Chromosome 6Pter-P24 Deletion Syndrome and Koolen-De Vries Syndrome. Further, an important paralog of the NAALADL2 gene is FOLH1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAALADL2
Applications Flow Cytometry, Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene Naaladl2
Gene Accession No. Q58DX5
Gene Alias 2810043G22Rik; EG635702; glutamate carboxypeptidase II-type non-peptidase homologue; Gm1021; Inactive N-acetylated-alpha-linked acidic dipeptidase-like protein 2; NAALADase L2; Naaladl2; N-acetylated alpha-linked acidic dipeptidase 2; N-acetylated alpha-linked acidic dipeptidase like 2; N-acetylated alpha-linked acidic dipeptidase-like 2; RGD1565147
Gene Symbols Naaladl2
Host Species Goat
Immunogen Mouse myeloma cell line NS0-derived recombinant human Naa LADase-like 2/Naa LADL2 Ser152-Asn795 (Met194Thr).
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 254827
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.