Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ NAB 2

Catalog No. 513110
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-311-0 10 mg
51-315-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 51-311-0 Supplier Tocris Bioscience™ Supplier No. 5131/10
This item is not returnable. View return policy

Protects against α-synuclein toxicity; promotes Rsp5/Nedd4-dependent endosomal transport

NAB 2 protects against α-synuclein toxicity. NAB 2 reverses the α-synuclein-induced pathological phenotype in Parkinson's disease cortical neurons. NAB 2 promotes E3 ubiquitin ligase Rsp5/Nedd4-dependent endosomal transport.

Specifications

Purity >98%
Quantity 10 mg
Product Type NAB 2
Molecular Weight (g/mol) 389.88
Inhibitors NAB 2

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.