Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAB1 Antibody (OTI1D2), HRP, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272867H
Description
NAB1 Monoclonal specifically detects NAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| NAB1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| NAB1 | |
| Recombinant protein fragment corresponding to amino acids 237-487 of human NAB1 (NP_005957) produced in E.coli. | |
| 0.1 mL | |
| 4664.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1D2 | |
| Western Blot, Immunohistochemistry | |
| EGR1 binding protein 1, EGR-1-binding protein 1, NGFI-A binding protein 1 (EGR1 binding protein 1), NGFI-A-binding protein 1, Transcriptional regulatory protein p54 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction