Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAB1 Monoclonal Antibody (OTI4G10), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA802913

Description
Acts as a transcriptional repressor for zinc finger transcription factors EGR1 and EGR2.Specifications
NAB1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q13506 | |
NAB1 | |
Human recombit protein fragment corresponding to amino acids 237-487 of human NAB1 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Western Blot | |
OTI4G10 | |
Unconjugated | |
NAB1 | |
NGFI-A binding protein 1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
4664 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction