Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540476
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540476 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540476 Supplier Invitrogen™ Supplier No. PA540476
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: ALRDWVTNPG LFNQPLTSLP VSSIPIYKLP EGSPTWLGIS CSSYERSSNA Sequence homology: Cow: 100%; Dog: 92%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 85%.

Acts as a transcriptional repressor for zinc finger transcription factors EGR1 and EGR2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAB1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene NAB1
Gene Accession No. Q13506
Gene Alias EGR1 binding protein 1; EGR-1-binding protein 1; Nab1; NGF1A-binding protein; Ngfi-A binding protein 1; NGFI-A binding protein 1 (EGR1 binding protein 1); NGFI-A-binding protein 1; Transcriptional regulatory protein p54
Gene Symbols NAB1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human NAB1 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4664
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.