Learn More
NAB1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31
Specifications
Antigen | NAB1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-156-4215
![]() |
Bethyl Laboratories, Inc
A303082AT |
20 μL |
Each of 1 for $151.31
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat and Golden hamster
Acts as a transcriptional repressor for zinc finger transcription factors EGR1 and EGR2.Specifications
NAB1 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
EGR-1-binding protein 1; EGR1 binding protein 1; NGFI-A-binding protein 1; transcriptional regulatory protein p54 | |
Between 460 and 487 | |
Primary | |
4° C | |
NAB1 |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q13506, Q61122 | |
17936, 4664 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.