Learn More
NAB1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303081AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Acts as a transcriptional repressor for zinc finger transcription factors EGR1 and EGR2.Specifications
NAB1 | |
Polyclonal | |
Unconjugated | |
NAB1 | |
EGR-1-binding protein 1; EGR1 binding protein 1; NGFI-A-binding protein 1; transcriptional regulatory protein p54 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4664 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q13506 | |
NAB1 | |
Between 410 and 460 | |
10 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.