Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Nab2 Antibody, DyLight 594, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP171869DL594
Description
Nab2 Polyclonal antibody specifically detects Nab2 in Human samples. It is validated for Western BlotSpecifications
Nab2 | |
Polyclonal | |
Western Blot | |
EGR-1-binding protein 2, MADEREGR1 binding protein 2, Melanoma-associated delayed early response protein, MGC75085, NGFI-A binding protein 2 (EGR1 binding protein 2), NGFI-A-binding protein 2, Protein MADER | |
The immunogen for this product maps to a region between residue 475 and 525 of human NGFI-A Binding Protein 2 using the numbering given in entry NP_005958.1 (GeneID 4665). | |
0.1 mL | |
Neuroscience, Transcription Factors and Regulators | |
4665 | |
Store at 4°C in the dark. | |
IgG |
Western Blot | |
DyLight 594 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction