Learn More
NAB2 Mouse anti-Human, Clone: OTI6D10, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF803954

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a member of the family of NGFI-A binding (NAB) proteins, which function in the nucleus to repress transcription induced by some members of the EGR (early growth response) family of transactivators. NAB proteins can homo- or hetero-multimerize with other EGR or NAB proteins through a conserved N-terminal domain, and repress transcription through two partially redundant C-terminal domains. Transcriptional repression by the encoded protein is mediated in part by interactions with the nucleosome remodeling and deactylase (NuRD) complex. Alternatively spliced transcript variants have been described, but their biological validity has not been determined.Specifications
NAB2 | |
Monoclonal | |
Unconjugated | |
NAB2 | |
MADER | |
Mouse | |
Affinity Chromatography | |
RUO | |
4665 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI6D10 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q15742 | |
NAB2 | |
Human recombit protein fragment corresponding to amino acids 178-442 of human NAB2 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.