Learn More
Invitrogen™ NABP1 Monoclonal Antibody (OTI6B10)

Description
NABP1 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | NABP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI6B10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | NABP1 |
| Gene Accession No. | Q96AH0 |
| Gene Alias | 4930434H03Rik; 4930442A21Rik; 4930488J04Rik; 4933440J18Rik; 5830411E10Rik; AI852561; hSSB2; Nabp1; Nbp1; nucleic acid binding protein 1; nucleic acid-binding protein 1; Obfc2a; oligonucleotide/oligosaccharide-binding fold containing 2A; oligonucleotide/oligosaccharide-binding fold-containing protein 2A; RGD1306658; Sensor of single-strand DNA complex subunit B2; sensor of ssDNA subunit B2; Single-stranded DNA-binding protein 2; SOSS complex subunit B2; SOSS-B2; Ssb2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.