Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | NAC1 |
---|---|
Dilution | Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
NAC1 Polyclonal specifically detects NAC1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
NAC1 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1 | |
NACC1 | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
Polyclonal | |
Rabbit | |
Cancer, Chromatin Research, Embryonic Stem Cell Markers, Stem Cell Markers, Transcription Factors and Regulators | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
112939 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:PGSYHNEEDEEEDGGEEGMDEQYRQICNMYTMYSMMNVGQTAEKVEALPEQVAPESRNRIRVRQDLASLPAELINQIGNRCH | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title