Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAC1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP299428100UL
Description
NAC1 Polyclonal antibody specifically detects NAC1 in Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
NAC1 | |
Polyclonal | |
Immunohistochemistry-Paraffin 1:500-1:2000 | |
BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human NAC1 | |
100 μg | |
Cancer, Chromatin Research, Stem Cell Markers, Transcription Factors and Regulators | |
112939 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
0.2 um filtered solution in PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction