Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAC1 Antibody (PCRP-NACC1-1A8), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308833APC
Description
NAC1 Monoclonal antibody specifically detects NAC1 in Human samples. It is validated for Flow Cytometry, Protein ArraySpecifications
NAC1 | |
Monoclonal | |
APC | |
BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1 | |
Recombinant full-length human NAC1 protein (Uniprot: Q96RE7) | |
100 μg | |
Cancer, Chromatin Research, Stem Cell Markers, Transcription Factors and Regulators | |
112939 | |
Store at 4°C in the dark. | |
IgG2a |
Flow Cytometry, Protein Array | |
PCRP-NACC1-1A8 | |
PBS | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction