Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
NAC1 Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298710PE
Description
NAC1 Polyclonal antibody specifically detects NAC1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
NAC1 | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
BEND8, BTB (POZ) domain containing 14B, BTB/POZ domain-containing protein 14B, BTBD14B, NAC1BEN domain containing 8, NAC-1FLJ37383, nucleus accumbens associated 1, BEN and BTB (POZ) domain containing, nucleus accumbens-associated protein 1, transcriptional repressor NAC1 | |
Produced in rabbits immunized with A synthetic peptide corresponding to the N-terminus of the human NAC1. | |
0.1 mL | |
Cancer, Chromatin Research, Stem Cell Markers, Transcription Factors and Regulators | |
112939 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction