Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAC1/BTBD14B Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501982073
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-2073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-2073 Supplier Bioss Supplier No. BS12247R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Members of the BTB/POZ family of transcriptional regulators, including BTBD14B, contain a conserved motif in the N-terminal region critical for protein-protein interactions and assembly of high molecular mass complexes (Korutla et al., 2002 [PubMed 11906783]).

Specifications

Antigen NAC1/BTBD14B
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene NACC1
Gene Accession No. Q96RE7
Gene Alias 2010001H03Rik; 4930511N13Rik; BEN domain containing 8; BEND8; BOS_7409; BTB (POZ) domain containing 14B; BTB/POZ domain-containing protein 14B; BTBD14B; BTBD30; FLJ37383; NAC1; NAC-1; Nacc1; nucleus accumbens associated 1; nucleus accumbens associated 1, BEN and BTB (POZ) domain containing; nucleus accumbens-1; Nucleus accumbens-associated protein 1; transcriptional repressor NAC1
Gene Symbols NACC1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human NAC1/BTBD14B.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112939
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.