Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAC1 Monoclonal Antibody (6H2)
GREENER_CHOICE

Catalog No. PIMA515727
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA515727 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA515727 Supplier Invitrogen™ Supplier No. MA515727
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA5-15727 targets NACC1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15727 immunogen is purified recombinant fragment of human NACC1 expressed in E. Coli. MA5-15727 detects NACC1 which has a predicted molecular weight of approximately 58kDa.

Members of the BTB/POZ family of transcriptional regulators, including BTBD14B, contain a conserved motif in the N-terminal region critical for protein-protein interactions and assembly of high molecular mass complexes (Korutla et al., 2002 [PubMed 11906783]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAC1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 6H2
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation ascites with 0.03% sodium azide
Gene NACC1
Gene Accession No. Q96RE7
Gene Alias 2010001H03Rik; 4930511N13Rik; BEN domain containing 8; BEND8; BTB (POZ) domain containing 14B; BTB/POZ domain-containing protein 14B; Btbd14b; BTBD30; FLJ37383; Nac1; NAC-1; Nacc1; nucleus accumbens associated 1; nucleus accumbens associated 1, BEN and BTB (POZ) domain containing; nucleus accumbens-1; nucleus accumbens-associated protein 1; nucleus accumbens-associated protein 1; LOW QUALITY PROTEIN: nucleus accumbens-associated protein 1; transcriptional repressor NAC1
Gene Symbols NACC1
Host Species Mouse
Immunogen Purified recombinant fragment of human NACC1 expressed in E. Coli.
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112939
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.