Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NAC1 Mouse anti-Human, Unconjugated, Clone: 9.27, MilliporeSigma™

Catalog No. MABC17131MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC17131MI 100 μg
MABC17132MI 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MABC17131MI Supplier MilliporeSigma Supplier No. MABC1713100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

NAC1 Monoclonal antibody specifically detects NAC1 in Human samples. It is validated for ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAC1
Applications ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 9.27
Conjugate Unconjugated
Gene Accession No. NP_443108
Gene Alias Nucleus accumbens-associated protein 1;BTB/POZ domain-containing protein 14B
Gene Symbols NACC1, NAC1, BTBD14B
Host Species Mouse
Immunogen GST-tagged full-length recombinant human NAC-1 protein.
Molecular Weight of Antigen 57.26 kDa calculated.
Purification Method Protein G Purified
Quantity 100 μg
Research Discipline Apoptosis and Cancer
Primary or Secondary Primary
Test Specificity Clone 9.27 is a mouse monoclonal antibody that detects Nucleus accumbens-associated protein 1 (NAC-1).
Target Species Human
Content And Storage Stable for 1 year at 2 to 8°C from date of receipt.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.