Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NAC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531474
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531474 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531474 Supplier Invitrogen™ Supplier No. PA531474
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse olfactory. Predicted reactivity: Mouse (93%), Rat (93%), Bovine (95%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Members of the BTB/POZ family of transcriptional regulators, including BTBD14B, contain a conserved motif in the N-terminal region critical for protein-protein interactions and assembly of high molecular mass complexes (Korutla et al., 2002 [PubMed 11906783]).
TRUSTED_SUSTAINABILITY

Specifications

Antigen NAC1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene NACC1
Gene Accession No. Q7TSZ8, Q96RE7
Gene Alias 2010001H03Rik; 4930511N13Rik; BEN domain containing 8; BEND8; BTB (POZ) domain containing 14B; BTB/POZ domain-containing protein 14B; Btbd14b; BTBD30; FLJ37383; Nac1; NAC-1; Nacc1; nucleus accumbens associated 1; nucleus accumbens associated 1, BEN and BTB (POZ) domain containing; nucleus accumbens-1; nucleus accumbens-associated protein 1; nucleus accumbens-associated protein 1; LOW QUALITY PROTEIN: nucleus accumbens-associated protein 1; transcriptional repressor NAC1
Gene Symbols NACC1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NAC1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 112939, 66830
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.