Learn More
Invitrogen™ NACA2 Monoclonal Antibody (4H6)

Description
Peptide Sequence: ASDAYIVFGE AKIQDLSQQA QLAAAEKFRV QGEAVGNIQE NTQTPTVQEE SEEEEVDETG VEVKDVKLVM SQANVSRAKA VRALKNNSND IVNAIMELTV.
Specifications
Specifications
| Antigen | NACA2 |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | 4H6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | NACA2 |
| Gene Accession No. | Q9H009 |
| Gene Alias | alpha-NAC protein; Alpha-NAC-like; ANAC; Hom s 2.01; hom s 2.01 protein; IgE autoantigen alpha-nascent polypeptide-associated complex; NACA2; NACAL; NAC-alpha-like; nascent polypeptide associated complex alpha subunit 2; nascent polypeptide-associated complex alpha subunit 2; nascent polypeptide-associated complex subunit alpha-2; Nascent polypeptide-associated complex subunit alpha-like; nascent polypeptide-associated complex, alpha polypeptide, 2; Prpk; Trp53rk |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.