Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ NACA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113481
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113481 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113481 Supplier Invitrogen™ Supplier No. PA5113481
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

NACA2 Polyclonal Antibody for Western Blot, ELISA

Prevents inappropriate targeting of non-secretory polypeptides to the endoplasmic reticulum (ER). Binds to nascent polypeptide chains as they emerge from the ribosome and blocks their interaction with the signal recognition particle (SRP), which normally targets nascent secretory peptides to the ER. Also reduces the inherent affinity of ribosomes for protein translocation sites in the ER membrane (M sites). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen NACA2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene NACA2
Gene Accession No. Q9H009
Gene Alias alpha-NAC protein; Alpha-NAC-like; ANAC; Hom s 2.01; hom s 2.01 protein; IgE autoantigen alpha-nascent polypeptide-associated complex; NACA2; NACAL; NAC-alpha-like; nascent polypeptide associated complex alpha subunit 2; nascent polypeptide-associated complex alpha subunit 2; nascent polypeptide-associated complex subunit alpha-2; Nascent polypeptide-associated complex subunit alpha-like; nascent polypeptide-associated complex, alpha polypeptide, 2; Prpk; Trp53rk
Gene Symbols NACA2
Host Species Rabbit
Immunogen Recombinant Human Nascent polypeptide-associated complex subunit alpha-2 protein (1-215AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 342538
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.